4-Bromo-3-(hydroxymethyl)benzonitrile 97%

Supplier: Ambeed
Danger

A394623-1G A394623-5G A394623-250MG
77168-254EA 115 USD
77168-254 77168-256 77892-812
4-Bromo-3-(hydroxymethyl)benzonitrile 97%
4-Bromo-3-(hydroxymethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD11858305
CAS Number: 905710-66-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR