1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,6-Di-i-propylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride, 1-(2,6-Diisopropylphenyl)-3-mesityl-4,5-dihydro-1H-imidazol-3-ium chloride

A394258-1G A394258-250MG A394258-5G
77195-872EA 92.7 USD
77195-872 77195-874 77195-876
1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride 98%
1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride

Precaución: for Research Use Only

Formula: C₂₄H₃₃ClN₂
MW: 384.992 g/mol
MDL Number: MFCD17018754
CAS Number: 866926-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR