1,1,3,3-Tetramethyl-2-thiourea 99%

Supplier: Ambeed
Warning

A393900-1000G A393900-100G A393900-25G A393900-500G A393900-5G
77195-862EA 51 USD
77195-862 77195-864 77195-866 77195-868 77195-870
1,1,3,3-Tetramethyl-2-thiourea 99%
1,1,3,3-Tetramethyl-2-thiourea

Precaución: for Research Use Only

Formula: C₅H₁₂N₂S
MDL Number: MFCD00008324
CAS Number: 2782-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR