Methyl 2-(2-hydroxy-4,5-dimethoxybenzamido)thiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A393100-100G A393100-25G A393100-5G
77195-850EA 393.46 USD
77195-850 77195-852 77195-854
Methyl 2-(2-hydroxy-4,5-dimethoxybenzamido)thiazole-4-carboxylate 97%
Methyl 2-(2-hydroxy-4,5-dimethoxybenzamido)thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O₆S
MW: 338.33 g/mol
MDL Number: MFCD26127250
CAS Number: 877997-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR