N-Methoxycarbonyl-L-tert-leucine 97%

Supplier: Ambeed
Warning

Synonyms: Moc-tert-Leu-OH, (S)-2-((Methoxycarbonyl)amino)-3,3-dimethylbutanoic acid

A393007-100G A393007-25G A393007-500G A393007-10G
77195-844EA 43 USD
77195-844 77195-846 77195-848 77572-190
N-Methoxycarbonyl-L-tert-leucine 97%
N-Methoxycarbonyl-L-tert-leucine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₄
MDL Number: MFCD07778455
CAS Number: 162537-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR