2-(4-Bromophenyl)-5-phenylthiophene 97%

Supplier: Ambeed
Warning

A393003-1G A393003-250MG A393003-5G
77195-838EA 56.65 USD
77195-838 77195-840 77195-842
2-(4-Bromophenyl)-5-phenylthiophene 97%
2-(4-Bromophenyl)-5-phenylthiophene

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrS
MW: 315.23 g/mol
MDL Number: MFCD21648452
CAS Number: 118621-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR