tert-Butyl 3-bromo-1H-pyrazole-1-carboxylate 98%

Supplier: Ambeed
Warning

A392915-100MG A392915-250MG A392915-1G A392915-5G
77168-232EA 45.32 USD
77168-232 77168-234 77551-166 77684-122
tert-Butyl 3-bromo-1H-pyrazole-1-carboxylate 98%
tert-Butyl 3-bromo-1H-pyrazole-1-carboxylate

Precaución: For research use only.

Formula: C₈H₁₁BrN₂O₂
MW: 247.089 g/mol
MDL Number: MFCD25976492
CAS Number: 1448855-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR