PLX-3397 98%

Supplier: Ambeed
Warning

Synonyms: Pexidartinib, 5-((5-Chloro-1H-pyrrolo[2,3-b]pyridin-3-yl)methyl)-N-((6-(trifluoromethyl)pyridin-3-yl)methyl)pyridin-2-amine

A392608-100MG A392608-1MG A392608-5MG A392608-250MG
77195-836EA 142.14 USD
77195-836 77616-686 77616-688 77746-787
PLX-3397 98%
PLX-3397

Precaución: for Research Use Only

Formula: C₂₀H₁₅ClF₃N₅
MW: 417.81 g/mol
MDL Number: MFCD28900745
CAS Number: 1029044-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR