Pentadecane-1,15-diol 98%

Supplier: Ambeed
Warning

Synonyms: Pentadecane-1,15-diol

A392596-100G A392596-1G A392596-5G A392596-25G
77195-830EA 576.8 USD
77195-830 77195-832 77195-834 77746-786
Pentadecane-1,15-diol 98%
Pentadecane-1,15-diol

Precaución: for Research Use Only

Formula: C₁₅H₃₂O₂
MW: 244.41 g/mol
MDL Number: MFCD00039633
CAS Number: 14722-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR