Ethyl 6-oxo-1,6-dihydropyrimidine-5-carboxylate 98%

Supplier: Ambeed
Warning

A391997-1G A391997-25G A391997-5G
77168-222EA 22.66 USD
77168-222 77168-224 77168-226
Ethyl 6-oxo-1,6-dihydropyrimidine-5-carboxylate 98%
Ethyl 6-oxo-1,6-dihydropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD00194922
CAS Number: 4786-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR