DBeQ 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-Dibenzylquinazoline-2,4-diamine, N2,N4-Dibenzylquinazoline-2,4-diamine

A391735-10MG A391735-1MG A391735-50MG A391735-5MG
77195-822EA 27.81 USD
77195-822 77195-824 77195-826 77195-828
DBeQ 98%
DBeQ

Precaución: for Research Use Only

Formula: C₂₂H₂₀N₄
MW: 340.43 g/mol
MDL Number: MFCD03691820
CAS Number: 177355-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR