2-Amino-5-hydroxy-6-methylpyrimidin-4(1H)-one 97%

Supplier: Ambeed
Warning

A391447-100MG A391447-1G A391447-250MG
77195-816EA 50.47 USD
77195-816 77195-818 77195-820
2-Amino-5-hydroxy-6-methylpyrimidin-4(1H)-one 97%
2-Amino-5-hydroxy-6-methylpyrimidin-4(1H)-one

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
MDL Number: MFCD19207675
CAS Number: 14405-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR