6-Chloro-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: Ambeed
Warning

A391269-1G A391269-250MG
77168-210EA 70.04 USD
77168-210 77168-212
6-Chloro-1,2,3,4-tetrahydroisoquinoline 97%
6-Chloro-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀ClN
MW: 167.6354 g/mol
MDL Number: MFCD06739156
CAS Number: 33537-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR