6-chloro-5-fluoroindole 96%

Supplier: Ambeed
Danger

Synonyms: 6-Chloro-5-fluoroindole

A391265-100G A391265-10G A391265-1G A391265-250MG A391265-25G A391265-5G
77195-806EA 2701.69 USD
77195-806 77195-808 77195-810 77195-812 77195-814 77616-682
6-chloro-5-fluoroindole 96%
6-chloro-5-fluoroindole

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MW: 169.583 g/mol
MDL Number: MFCD01074496
CAS Number: 122509-72-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR