3,3'-(Ethane-1,2-diyl)bis(1H-pyrrole-2,5-dione) 98%

Supplier: Ambeed
Warning

A390889-100MG A390889-1G A390889-250MG
77195-800EA 24.72 USD
77195-800 77195-802 77195-804
3,3'-(Ethane-1,2-diyl)bis(1H-pyrrole-2,5-dione) 98%
3,3'-(Ethane-1,2-diyl)bis(1H-pyrrole-2,5-dione)

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₄
MW: 220.18 g/mol
CAS Number: 1823773-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR