tert-Butyl (4-bromocyclohexyl)carbamate 96%

Supplier: Ambeed
Warning

A390262-100MG A390262-1G A390262-5G
77168-204EA 80 USD
77168-204 77168-206 77168-208
tert-Butyl (4-bromocyclohexyl)carbamate 96%
tert-Butyl (4-bromocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₀BrNO₂
MDL Number: MFCD21097795
CAS Number: 1353963-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR