Piperidin-1-ylacetic acid 98%

Supplier: Ambeed
Danger

Synonyms: 1-Piperidineacetic Acid

A390197-1G A390197-5G
77168-200EA 28.84 USD
77168-200 77168-202
Piperidin-1-ylacetic acid 98%
Piperidin-1-ylacetic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD00461042
CAS Number: 3235-67-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR