3-Bromopicolinamide 97%

Supplier: Ambeed
Warning

A389746-1G A389746-250MG A389746-5G
77195-786EA 73.13 USD
77195-786 77195-788 77195-790
3-Bromopicolinamide 97%
3-Bromopicolinamide

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O
MW: 201.02 g/mol
MDL Number: MFCD11043941
CAS Number: 36057-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR