4-(Bromomethyl)tetrahydropyran 95%

Supplier: Ambeed
Danger

A387739-10G A387739-1G A387739-25G A387739-5G
77168-150EA 55.62 USD
77168-150 77168-152 77168-154 77168-156
4-(Bromomethyl)tetrahydropyran 95%
4-(Bromomethyl)tetrahydropyran

Precaución: for Research Use Only

Formula: C₆H₁₁BrO
MW: 179.0549 g/mol
MDL Number: MFCD03788561
CAS Number: 125552-89-8
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR