2-Allyl-D-glycine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: D-Allylglycine, H-D-Allyl-Gly-OH, H-D-Gly(Allyl)-OH

A387684-1G A387684-250MG A387684-5G A387684-10G A387684-100G A387684-25G
77168-144EA 42 USD
77168-144 77168-146 77168-148 77572-168 77684-116 77684-117
2-Allyl-D-glycine 97%, Ambeed.Inc
2-Allyl-D-glycine

Precaución: For research use only.

Formula: C₅H₉NO₂
MW: 115.1 g/mol
MDL Number: MFCD00063104
CAS Number: 54594-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR