Ethyl 2-amino-4,5-difluorobenzoate 97%

Supplier: Ambeed
Warning

A387307-5G A387307-1G
77195-774EA 96.82 USD
77195-774 77572-160
Ethyl 2-amino-4,5-difluorobenzoate 97%
Ethyl 2-amino-4,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₉F₂NO₂
MW: 201.17 g/mol
MDL Number: MFCD11226323
CAS Number: 864293-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR