1-(6-Bromoimidazo[1,2-a]pyridin-3-yl)ethanone 97%

Supplier: Ambeed
Warning

A387168-1G A387168-250MG A387168-5G
77195-766EA 74.16 USD
77195-766 77195-768 77195-770
1-(6-Bromoimidazo[1,2-a]pyridin-3-yl)ethanone 97%
1-(6-Bromoimidazo[1,2-a]pyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MW: 239.06 g/mol
MDL Number: MFCD20260166
CAS Number: 30493-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR