4-(Di(1H-indol-3-yl)methyl)phenol 97%

Supplier: Ambeed
Warning

Synonyms: DIM-C-pPhOH

A386994-10MG A386994-1MG A386994-25MG A386994-5MG A386994-1G
77195-758EA 13.39 USD
77195-758 77195-760 77195-762 77195-764 77746-777
4-(Di(1H-indol-3-yl)methyl)phenol 97%
4-(Di(1H-indol-3-yl)methyl)phenol

Precaución: for Research Use Only

Formula: C₂₃H₁₈N₂O
MW: 338.40 g/mol
MDL Number: MFCD03723253
CAS Number: 151358-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR