1,1'-Diethylferrocene 97%

Supplier: Ambeed
Warning

Synonyms: Bis(ethylcyclopentadienyl)iron, 1,1'-Diethyl Ferrocene

A386773-1G A386773-5G
77195-754EA 18.54 USD
77195-754 77195-756
1,1'-Diethylferrocene 97%
1,1'-Diethylferrocene

Precaución: for Research Use Only

Formula: C₁₄H₁₀Fe
MW: 234.07 g/mol
MDL Number: MFCD01862455
CAS Number: 1273-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR