4-(3-Methoxybenzyl)piperidine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A386719-250MG
A386719-1G
77168-142EA
49.44
USD
77168-142
77684-115
4-(3-Methoxybenzyl)piperidine hydrochloride 95%
4-(3-Methoxybenzyl)piperidine hydrochloride
Precaución: For research use only.
Formula:
C₁₃H₂₀ClNO MW: 241.75 g/mol |
MDL Number:
MFCD08669672 CAS Number: 149986-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...