N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)-3-fluorophenyl)-4-ethoxy-1-(4-fluoro-2-methylphenyl)-1H-pyrazole-3-carboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: LDC1267

A386651-10MG A386651-1MG A386651-25MG A386651-5MG
77168-134EA 38.11 USD
77168-134 77168-136 77168-138 77168-140
N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)-3-fluorophenyl)-4-ethoxy-1-(4-fluoro-2-methylphenyl)-1H-pyrazole-3-carboxamide 98+%
N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)-3-fluorophenyl)-4-ethoxy-1-(4-fluoro-2-methylphenyl)-1H-pyrazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₃₀H₂₆F₂N₄O₅
MW: 560.55 g/mol
MDL Number: MFCD28385861
CAS Number: 1361030-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR