AZD-2014 ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A386287-10MG A386287-250MG A386287-5MG A386287-50MG
77168-122EA 69 USD
77168-122 77168-124 77168-126 77168-128 77684-111
AZD-2014 ≥98%, Ambeed.Inc
AZD-2014

Precaución: For research use only.

Formula: C₂₅H₃₀N₆O₃
MDL Number: MFCD22628784
CAS Number: 1009298-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR