tert-Butyl-N-(2-hydroxypropyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-(2-Hydroxypropyl)carbamic Acid tert-Butyl Ester, 1-(tert-Butoxycarbonylamino)-2-propanol, 1-(Boc-amino)-2-propanol, tert-Butyl (2-hydroxypropyl)carbamate

A385329-100G A385329-10G A385329-250MG A385329-25G A385329-5G
77195-738EA 847 USD
77195-738 77195-740 77195-742 77195-744 77195-746
tert-Butyl-N-(2-hydroxypropyl)carbamate 95%
tert-Butyl-N-(2-hydroxypropyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃
MDL Number: MFCD02093569
CAS Number: 95656-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR