Bicyclohexane-4,4'-dione monoethylene ketal 98%

Supplier: Ambeed
Warning

Synonyms: 8-(4-Oxocyclohexyl)-1,4-dioxaspiro[4.5]decane, Bicyclohexane-4,4-dione Monoethylene Ketal

A385025-100G A385025-25G A385025-5G
77195-732EA 92.7 USD
77195-732 77195-734 77195-736
Bicyclohexane-4,4'-dione monoethylene ketal 98%
Bicyclohexane-4,4'-dione monoethylene ketal

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₃
MW: 238.33 g/mol
MDL Number: MFCD00809704
CAS Number: 56309-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR