Methyl 2-(2-methoxyphenyl)-2-oxoacetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methoxy-α-oxobenzeneacetate

A385014-100MG A385014-1G A385014-250MG
77168-110EA 63.86 USD
77168-110 77168-112 77168-114
Methyl 2-(2-methoxyphenyl)-2-oxoacetate 95%
Methyl 2-(2-methoxyphenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD11617598
CAS Number: 5180-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR