1,3,8-Triazaspiro[4.5]decane-2,4-dione 95%

Supplier: Ambeed
Warning

A385013-1G A385013-250MG A385013-5G
77195-726EA 16 USD
77195-726 77195-728 77195-730
1,3,8-Triazaspiro[4.5]decane-2,4-dione 95%
1,3,8-Triazaspiro[4.5]decane-2,4-dione

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MDL Number: MFCD06660736
CAS Number: 13625-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR