1-Heptyl isothiocyanate 98%

Supplier: Ambeed
Danger

Synonyms: Heptylisothiocyanate

A384887-25G A384887-5G
77195-722EA 41.2 USD
77195-722 77195-724
1-Heptyl isothiocyanate 98%
1-Heptyl isothiocyanate

Precaución: for Research Use Only

Formula: C₈H₁₅NS
MW: 157.28 g/mol
MDL Number: MFCD00041138
CAS Number: 4426-83-9
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR