1,4-Butanediol dimethacrylate 95% stabilized

Supplier: Ambeed
Warning

Synonyms: Tetramethylene glycol dimethacrylate, TMA

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A384847-100G A384847-25G
77195-716EA 24.72 USD
77195-716 77195-718 77195-720
1,4-Butanediol dimethacrylate 95% stabilized
1,4-Butanediol dimethacrylate

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₄
MW: 226.27 g/mol
MDL Number: MFCD00008594
CAS Number: 2082-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR