Methyl chroman-3-carboxylate 96%

Supplier: Ambeed
Warning

A384785-1G A384785-250MG A384785-5G A384785-100MG
77168-104EA 66.95 USD
77168-104 77168-106 77594-160 77613-146
Methyl chroman-3-carboxylate 96%
Methyl chroman-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
MDL Number: MFCD04114625
CAS Number: 68281-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR