Diglycidyl-1,2-cyclohexanedicarboxylate 90% (by GC)

Supplier: Ambeed
Warning

Synonyms: 1,2-Cyclohexanedicarboxylic Acid Diglycidyl Ester, Bis(oxiran-2-ylmethyl) cyclohexane-1,2-dicarboxylate

A384508-100G A384508-25G A384508-500G A384508-1000G
77195-710EA 28.84 USD
77195-710 77195-712 77195-714 77572-142
Diglycidyl-1,2-cyclohexanedicarboxylate 90% (by GC)
Diglycidyl-1,2-cyclohexanedicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₆
MW: 284.31 g/mol
MDL Number: MFCD00128914
CAS Number: 5493-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR