Geniposide 99+%

Supplier: Ambeed
Warning

Synonyms: (1S,4aS,7aS)-Methyl 7-(hydroxymethyl)-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate, (1S)-1alpha-(beta-D-Glucopyranosyloxy)-7-(hydroxymethyl)-1,4a alpha,5,7a alpha-tetrahydrocyclopenta[c]pyran-4-carboxylic Acid M, Methyl (1S)-1alpha-(beta-D-Glucopyranosyloxy)-7-(hydroxymethyl)-1,4aalpha,5,7aalpha-tetrahydrocyclopenta[c]pyran-4-carboxylate

A383596-100MG A383596-1MG A383596-50MG A383596-5G A383596-25G A383596-250MG
77195-694EA 8.24 USD
77195-694 77195-696 77195-698 77195-700 77616-666 77746-771
Geniposide 99+%
Geniposide

Precaución: for Research Use Only

Formula: C₁₇H₂₄O₁₀
MW: 388.37 g/mol
MDL Number: MFCD16036219
CAS Number: 24512-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR