(2,4-Difluoro-3-propoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A383357-1G A383357-250MG A383357-5G
77195-688EA 87.55 USD
77195-688 77195-690 77195-692
(2,4-Difluoro-3-propoxyphenyl)boronic acid 97%
(2,4-Difluoro-3-propoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BF₂O₃
MW: 215.98 g/mol
MDL Number: MFCD19981527
CAS Number: 2096329-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR