3-Oxohexahydro-1H-pyrrolizine-7a-carbonitrile 97%

Supplier: Ambeed
Warning

A382958-100MG A382958-250MG
77195-682EA 86.52 USD
77195-682 77195-684
3-Oxohexahydro-1H-pyrrolizine-7a-carbonitrile 97%
3-Oxohexahydro-1H-pyrrolizine-7a-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.17 g/mol
MDL Number: MFCD28960645
CAS Number: 113089-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR