2-Methyl-2-(pyridin-2-yl)propan-1-amine 97%

Supplier: Ambeed
Danger

A382507-1G A382507-250MG
77195-676EA 1619.16 USD
77195-676 77195-678
2-Methyl-2-(pyridin-2-yl)propan-1-amine 97%
2-Methyl-2-(pyridin-2-yl)propan-1-amine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MW: 150.22 g/mol
MDL Number: MFCD11100803
CAS Number: 199296-39-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR