2,6-Bis(dodecyloxy)benzaldehyde 97%

Supplier: Ambeed
Warning

A382131-1G A382131-250MG A382131-5G
77168-074EA 88.58 USD
77168-074 77168-076 77168-078
2,6-Bis(dodecyloxy)benzaldehyde 97%
2,6-Bis(dodecyloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₃₁H₅₄O₃
MDL Number: MFCD25372091
CAS Number: 1198467-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR