(3-(Isopropoxymethyl)phenyl)boronic acid 98+%

Supplier: Ambeed
Warning

A381418-100MG A381418-1G A381418-250MG
77168-066EA 36.05 USD
77168-066 77168-068 77168-070
(3-(Isopropoxymethyl)phenyl)boronic acid 98+%
(3-(Isopropoxymethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.03 g/mol
MDL Number: MFCD14687288
CAS Number: 1333122-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR