Boc-L-Cys-OH 98%

Supplier: Ambeed
Danger

Synonyms: N-Boc-L-cysteine, (R)-2-((tert-Butoxycarbonyl)amino)-3-mercaptopropanoic acid

A381114-1G A381114-250MG A381114-25G A381114-5G
77195-660EA 30.9 USD
77195-660 77195-662 77195-664 77195-666
Boc-L-Cys-OH 98%
Boc-L-Cys-OH

Precaución: for Research Use Only

Formula: C₈H₁₅NO₄S
MW: 221.3 g/mol
MDL Number: MFCD00065565
CAS Number: 20887-95-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR