6-Nitro-o-toluidine 98%

Supplier: Ambeed
Danger

A380761-10G A380761-25G
77195-656EA 5 USD
77195-656 77616-656
6-Nitro-o-toluidine 98%
6-Nitro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD00007744
CAS Number: 570-24-1
UN: 2660
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR