1,6-Anhydro-β-D-glucose 98%

Supplier: Ambeed
Warning

A380273-1G A380273-250MG A380273-25G A380273-5G A380273-100G
77195-634EA 15 USD
77195-634 77195-636 77195-638 77195-640 77616-654
1,6-Anhydro-β-D-glucose 98%
1,6-Anhydro-beta-D-glucose

Precaución: for Research Use Only

Formula: C₆H₁₀O₅
MDL Number: MFCD00063248
CAS Number: 498-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR