1,2,4,5-Tetrakis(bromomethyl)benzene 95%

Supplier: Ambeed
Danger

A380180-1G A380180-5G
77195-630EA 10.3 USD
77195-630 77195-632
1,2,4,5-Tetrakis(bromomethyl)benzene 95%
1,2,4,5-Tetrakis(bromomethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀Br₄
MW: 449.8 g/mol
MDL Number: MFCD00009664
CAS Number: 15442-91-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR