Methyl 6-(aminomethyl)picolinate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A379801-100MG
A379801-1G
A379801-250MG
A379801-5G
77168-046EA
86.52
USD
77168-046
77168-048
77168-050
77168-052
Methyl 6-(aminomethyl)picolinate hydrochloride 95%
Methyl 6-(aminomethyl)picolinate hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₁ClN₂O₂ MW: 202.63 g/mol |
MDL Number:
MFCD16660686 CAS Number: 171670-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...