6-Bromoimidazo[1,2-a]pyridin-8-ol dihydrochloride 97%

Supplier: Ambeed
Warning

A379723-100MG A379723-1G A379723-250MG
77195-620EA 23.69 USD
77195-620 77195-622 77195-624
6-Bromoimidazo[1,2-a]pyridin-8-ol dihydrochloride 97%
6-Bromoimidazo[1,2-a]pyridin-8-ol dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₇BrCl₂N₂O
MW: 285.95 g/mol
MDL Number: MFCD28142911
CAS Number: 1379358-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR