(S)-2,4-Dichloro-alpha-(chloromethyl)benzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-alpha-(Chloromethyl)-2,4-dichlorobenzyl alcohol, (S)-2-Chloro-1-(2,4-dichlorophenyl)ethanol

A379620-100G A379620-25G A379620-500G A379620-5G
77195-612EA 46.35 USD
77195-612 77195-614 77195-616 77195-618
(S)-2,4-Dichloro-alpha-(chloromethyl)benzyl alcohol 97%
(S)-2,4-Dichloro-alpha-(chloromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₇Cl₃O
MW: 225.49 g/mol
MDL Number: MFCD07776655
CAS Number: 126534-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR