2-Chloro-5-propylpyrimidine 97%

Supplier: Ambeed
Warning

A378645-100MG A378645-1G A378645-250MG
77195-590EA 18 USD
77195-590 77195-592 77195-594
2-Chloro-5-propylpyrimidine 97%
2-Chloro-5-propylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MW: 156.61
MDL Number: MFCD00799403
CAS Number: 219555-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR