1,4-Dibromobenzene 98%

Supplier: Ambeed
Warning

A378578-1000G A378578-100G A378578-25G A378578-500G
77195-582EA 30.9 USD
77195-582 77195-584 77195-586 77195-588
1,4-Dibromobenzene 98%
1,4-Dibromobenzene

Precaución: for Research Use Only

Formula: C₆H₄Br₂
MW: 235.91 g/mol
MDL Number: MFCD00000089
CAS Number: 106-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR